Assay Details
Target Gene Details
Entrez Gene ID: | 64151 |
Gene Name: | non-SMC condensin I complex subunit G |
Gene Aliases: |
CAPG, CHCG, NY-MEL-3, YCG1 |
Location: |
Chr.4:17810813-17844865 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCAPG | NM_022346.4 | NP_071741.2 | ||
| NR_073124.1 | ||||
| XM_011513876.2 | XP_011512178.1 | |||
| XM_017008543.1 | XP_016864032.1 | |||
| XM_017008544.1 | XP_016864033.1 | |||
| AB013299.1 | BAB21557.1 | |||
| AF235023.1 | AAG30732.1 | |||
| AF331796.1 | AAG49627.1 | |||
| AK022512.1 | BAB14069.1 | |||
| AK023147.1 | BAB14429.1 | |||
| AK027511.1 | BAB55165.1 | |||
| BC000827.1 | AAH00827.1 | |||
| BC068467.1 | AAH68467.1 | |||
| BC101476.1 | AAI01477.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2425595 | Chr.4:17828236 - 17829850 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map