Assay Details
Target Gene Details
Entrez Gene ID: | 2788 |
Gene Name: | G protein subunit gamma 7 |
Gene Aliases: |
- |
Location: |
Chr.19:2511219-2702748 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNG7 | NM_052847.2 | 5 | 3526 | NP_443079.1 |
| XM_017026606.1 | 5 | 3635 | XP_016882095.1 | |
| AK024465.1 | 2 | 3115 | BAB15755.1 | |
| AK123863.1 | 1 | 2530 | ||
| BC053630.1 | 5 | 3443 | AAH53630.1 |
Target Gene Details
Entrez Gene ID: | 101929097 |
Gene Name: | uncharacterized LOC101929097 |
Gene Aliases: |
- |
Location: |
Chr.19:2511219-2513571 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929097 | XM_005259695.2 | 3 | 593 | XP_005259752.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064930 | Chr.19:2380093 - 2592252 on Build GRCh38 | Gain |
|
| esv2762019 | Chr.19:2503630 - 2519855 on Build GRCh38 | Gain |
|
| nsv953944 | Chr.19:2283702 - 2525802 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map