Assay Details
Target Gene Details
Entrez Gene ID: | 1390 |
Gene Name: | cAMP responsive element modulator |
Gene Aliases: |
CREM-2, ICER, hCREM-2 |
Location: |
Chr.10:35126841-35212958 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREM | NM_001267562.1 | 1 | 140 | NP_001254491.1 |
| NM_001267563.1 | 1 | 140 | NP_001254492.1 | |
| NM_001881.3 | 1 | 140 | NP_001872.3 | |
| NM_183013.2 | 1 | 140 | NP_898831.1 | |
| NR_051973.1 | 1 | 140 | ||
| NR_051974.1 | 1 | 140 | ||
| NR_051975.1 | 1 | 140 | ||
| XM_017015716.1 | 1 | 89 | XP_016871205.1 | |
| XM_017015717.1 | XP_016871206.1 | |||
| XM_017015719.1 | XP_016871208.1 | |||
| XM_017015723.1 | XP_016871212.1 | |||
| XM_017015724.1 | 1 | 140 | XP_016871213.1 | |
| XM_017015727.1 | XP_016871216.1 | |||
| XM_017015728.1 | 1 | 140 | XP_016871217.1 | |
| XM_017015729.1 | XP_016871218.1 | |||
| XM_017015737.1 | XP_016871226.1 | |||
| BC017117.1 | AAH17117.1 | |||
| BC066342.1 | AAH66342.1 | |||
| BG573680.1 | 1 | 109 | ||
| CB996061.1 | ||||
| CN367927.1 | ||||
| DA756152.1 | 1 | 140 | ||
| DR760671.1 |
Target Gene Details
Entrez Gene ID: | 8453 |
Gene Name: | cullin 2 |
Gene Aliases: |
- |
Location: |
Chr.10:35008551-35126899 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CUL2 | XM_011519745.1 | XP_011518047.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039051 | Chr.10:35049745 - 35203997 on Build GRCh38 | Gain |
|
| dgv706n100 | Chr.10:34881487 - 35230186 on Build GRCh38 | Gain |
|
| nsv550398 | Chr.10:34885707 - 35271661 on Build GRCh38 | Gain |
|
| esv3576372 | Chr.10:35088547 - 35160753 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map