Assay Details
Target Gene Details
Entrez Gene ID: | 9970 |
Gene Name: | nuclear receptor subfamily 1 group I member 3 |
Gene Aliases: |
CAR, CAR1, MB67 |
Location: |
Chr.1:161229666-161238623 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
More Information
Set Membership: |
|

Genomic Map