Assay Details
Target Gene Details
Entrez Gene ID: | 100506023 |
Gene Name: | uncharacterized LOC100506023 |
Gene Aliases: |
- |
Location: |
Chr.1:173235060-173477155 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506023 | NR_037845.1 | |||
| AK127238.1 |
Target Gene Details
Entrez Gene ID: | 101928673 |
Gene Name: | uncharacterized LOC101928673 |
Gene Aliases: |
- |
Location: |
Chr.1:173417789-173461362 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928673 | NR_125960.1 | 2 | 1402 | |
| AK130460.1 | 2 | 1403 | BAC85357.1 | |
| BU735514.1 | 1 | 434 |
Target Gene Details
Entrez Gene ID: | 7292 |
Gene Name: | tumor necrosis factor superfamily member 4 |
Gene Aliases: |
CD134L, CD252, GP34, OX-40L, OX4OL, TNLG2B, TXGP1 |
Location: |
Chr.1:173183729-173462208 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFSF4 | XM_017002229.1 | XP_016857718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831915 | Chr.1:173299285 - 173513018 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map