Assay Details
Target Gene Details
Entrez Gene ID: | 8909 |
Gene Name: | endonuclease, poly(U) specific |
Gene Aliases: |
P11, PP11, PRSS26 |
Location: |
Chr.12:47709734-47725572 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENDOU | NM_001172439.1 | 6 | 878 | NP_001165910.1 |
| NM_001172440.1 | 4 | 689 | NP_001165911.1 | |
| NM_006025.3 | 5 | 755 | NP_006016.1 | |
| AK075446.1 | 6 | 842 | ||
| AK295963.1 | 3 | 477 | ||
| AK300169.1 | 4 | 651 | ||
| AK302308.1 | 3 | 469 | ||
| AK314688.1 | 5 | 625 | ||
| AX772808.1 | 6 | 830 | CAE06253.1 | |
| BC069715.1 | 5 | 634 | AAH69715.1 | |
| BC074763.2 | 5 | 640 | AAH74763.1 | |
| BC111782.1 | 5 | 729 | AAI11783.1 | |
| M32402.1 | 5 | 750 | AAA36464.1 | |
| M36109.1 | 5 | 750 | AAA36465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751101 | Chr.12:47547075 - 47733102 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map