Assay Details
Target Gene Details
Entrez Gene ID: | 571 |
Gene Name: | BTB domain and CNC homolog 1 |
Gene Aliases: |
BACH-1, BTBD24 |
Location: |
Chr.21:29298795-29361896 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BACH1 | NM_001186.3 | NP_001177.1 | ||
| NM_206866.2 | NP_996749.1 | |||
| NR_027655.2 | ||||
| AA402241.1 | ||||
| AB002803.1 | BAA24932.1 | |||
| AF026199.1 | AAB84100.1 | |||
| AK291021.1 | ||||
| AK313791.1 | ||||
| BC063307.1 | AAH63307.1 | |||
| CR450303.1 | CAG29299.1 | |||
| DA855751.1 | ||||
| HY028322.1 |
More Information
Set Membership: |
|

Genomic Map