Assay Details
Target Gene Details
Entrez Gene ID: | 10153 |
Gene Name: | CCAAT/enhancer binding protein zeta |
Gene Aliases: |
CBF, CBF2, HSP-CBF, NOC1 |
Location: |
Chr.2:37201629-37231597 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPZ | NM_005760.2 | NP_005751.2 | ||
| AK312322.1 | ||||
| BC034475.2 | AAH34475.1 | |||
| M37197.1 | AAA51924.1 |
Target Gene Details
Entrez Gene ID: | 100505876 |
Gene Name: | CEBPZ opposite strand |
Gene Aliases: |
CEBPZ-AS1 |
Location: |
Chr.2:37196451-37216193 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEBPZOS | NM_001322373.1 | 6 | 1446 | NP_001309302.1 |
| NM_001322374.1 | 5 | 1267 | NP_001309303.1 | |
| NM_001322375.1 | NP_001309304.1 | |||
| NM_001322376.1 | 5 | 1263 | NP_001309305.1 | |
| NR_136316.1 | ||||
| XM_017003106.1 | 6 | 1475 | XP_016858595.1 | |
| XM_017003107.1 | 5 | 1296 | XP_016858596.1 | |
| XM_017003108.1 | XP_016858597.1 | |||
| XM_017003109.1 | XP_016858598.1 | |||
| AK001814.1 | 6 | 1180 | ||
| BM838731.1 |
More Information
Set Membership: |
|

Genomic Map