Assay Details
Target Gene Details
Entrez Gene ID: | 493860 |
Gene Name: | coiled-coil domain containing 73 |
Gene Aliases: |
NY-SAR-79 |
Location: |
Chr.11:32602080-32794658 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC73 | NM_001008391.3 | 16 | 1984 | NP_001008392.2 |
| XM_011520139.2 | 15 | 1845 | XP_011518441.1 | |
| AK128159.1 | 7 | 1203 | BAC87304.1 | |
| AK131470.1 | 16 | 1968 | BAD18615.1 | |
| AY211922.1 | 1 | 78 | AAO65175.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs117983166] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map