Assay Details
Target Gene Details
Entrez Gene ID: | 100288866 |
Gene Name: | uncharacterized LOC100288866 |
Gene Aliases: |
- |
Location: |
Chr.17:49505657-49574064 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100288866 | NR_103773.1 | 3 | 458 | |
| AK303671.1 | 3 | 458 |
Target Gene Details
Entrez Gene ID: | 4804 |
Gene Name: | nerve growth factor receptor |
Gene Aliases: |
CD271, Gp80-LNGFR, TNFRSF16, p75(NTR), p75NTR |
Location: |
Chr.17:49495293-49515020 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NGFR | NM_002507.3 | 3 | 455 | NP_002498.1 |
| AK303278.1 | 3 | 523 | ||
| AK313654.1 | 3 | 455 | ||
| BC050309.1 | 3 | 455 | AAH50309.1 | |
| DA115173.1 | 3 | 455 | ||
| M14764.1 | 3 | 443 | AAB59544.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833473 | Chr.17:49383665 - 49545926 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map