Assay Details
Target Gene Details
Entrez Gene ID: | 11218 |
Gene Name: | DEAD-box helicase 20 |
Gene Aliases: |
DP103, GEMIN3 |
Location: |
Chr.1:111755568-111767577 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX20 | NM_007204.4 | 11 | 3276 | NP_009135.4 |
| AF155803.1 | 11 | 2924 | AAK56799.1 | |
| AL133598.1 | 3 | 4100 | CAB63734.2 | |
| AY007146.1 | 1 | 879 | AAG02003.1 | |
| BC034953.2 | 11 | 3247 | AAH34953.1 |
More Information
Set Membership: |
|

Genomic Map