Assay Details
Target Gene Details
Entrez Gene ID: | 2675 |
Gene Name: | GDNF family receptor alpha 2 |
Gene Aliases: |
GDNFRB, NRTNR-ALPHA, NTNRA, RETL2, TRNR2 |
Location: |
Chr.8:21690403-21789296 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GFRA2 | NM_001165038.1 | NP_001158510.1 | ||
| NM_001165039.1 | NP_001158511.1 | |||
| NM_001495.4 | NP_001486.4 | |||
| XM_006716327.3 | XP_006716390.1 | |||
| XM_011544484.2 | XP_011542786.1 | |||
| AF002700.1 | AAC52036.1 | |||
| AK299623.1 | ||||
| AY326396.1 | AAP88378.1 | |||
| AY941828.1 | AAX46325.1 | |||
| BC041688.1 | AAH41688.1 | |||
| U93703.1 | AAB61922.1 | |||
| U97145.1 | AAC51647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891375 | Chr.8:21615265 - 21795198 on Build GRCh38 | Gain |
|
| esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map