Assay Details
Target Gene Details
Entrez Gene ID: | 22941 |
Gene Name: | SH3 and multiple ankyrin repeat domains 2 |
Gene Aliases: |
AUTS17, CORTBP1, CTTNBP1, ProSAP1, SHANK, SPANK-3 |
Location: |
Chr.11:70467856-71252724 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHANK2 | NM_012309.4 | NP_036441.2 | ||
| XM_017017387.1 | XP_016872876.1 | |||
| XM_017017388.1 | XP_016872877.1 | |||
| XM_017017389.1 | XP_016872878.1 | |||
| AB208025.1 | ||||
| DQ152234.1 | AAZ77790.1 |
Target Gene Details
Entrez Gene ID: | 220070 |
Gene Name: | SHANK2 antisense RNA 3 |
Gene Aliases: |
C11orf76 |
Location: |
Chr.11:70862790-70865115 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHANK2-AS3 | NR_073536.1 | |||
| BC004224.2 | AAH04224.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv381 | Chr.11:70850842 - 70884302 on Build GRCh38 | Insertion |
|
| nsv555399 | Chr.11:70851929 - 71158475 on Build GRCh38 | Loss |
|
| esv993927 | Chr.11:70861407 - 70874735 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map