Assay Details
Target Gene Details
Entrez Gene ID: | 148641 |
Gene Name: | solute carrier family 35 member F3 |
Gene Aliases: |
- |
Location: |
Chr.1:233904712-234324518 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35F3 | NM_173508.3 | 1 | 328 | NP_775779.1 |
| AK095031.1 | 1 | 109 | BAC04479.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832892 | Chr.1:233784001 - 233964848 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map