Assay Details
Target Gene Details
Entrez Gene ID: | 6483 |
Gene Name: | ST3 beta-galactoside alpha-2,3-sialyltransferase 2 |
Gene Aliases: |
Gal-NAc6S, SIAT4B, ST3GALII, ST3GalA.2 |
Location: |
Chr.16:70379435-70439088 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST3GAL2 | NM_006927.3 | 2 | 968 | NP_008858.1 |
| AK127322.1 | 1 | 1933 | ||
| AK313020.1 | 1 | 99 | ||
| U63090.1 | 1 | 504 | AAB40389.1 | |
| X96667.1 | 1 | 121 | CAA65447.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833272 | Chr.16:70335866 - 70535820 on Build GRCh38 | Loss |
|
| esv3892881 | Chr.16:70379334 - 70791080 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map