Assay Details
Target Gene Details
Entrez Gene ID: | 2918 |
Gene Name: | glutamate metabotropic receptor 8 |
Gene Aliases: |
GLUR8, GPRC1H, MGLUR8, mGlu8 |
Location: |
Chr.7:126438598-127253294 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRM8 | NM_000845.2 | 10 | 3251 | NP_000836.2 |
| NM_001127323.1 | 11 | 3552 | NP_001120795.1 | |
| NR_028041.1 | 11 | 3322 | ||
| XM_006715938.3 | 11 | 3571 | XP_006716001.1 | |
| XM_011516091.1 | 11 | 3749 | XP_011514393.1 | |
| XM_011516092.2 | 11 | 3804 | XP_011514394.1 | |
| XM_011516094.1 | 11 | 2550 | XP_011514396.1 | |
| XM_011516095.2 | 11 | 2683 | XP_011514397.1 | |
| XM_011516101.2 | 10 | 2555 | XP_011514403.1 | |
| XM_011516102.2 | 11 | 2580 | XP_011514404.1 | |
| XM_011516103.2 | 11 | 2712 | XP_011514405.1 | |
| XM_011516108.2 | 6 | 2654 | XP_011514410.1 | |
| XM_017012074.1 | 11 | 3350 | XP_016867563.1 | |
| XM_017012075.1 | 11 | 2701 | XP_016867564.1 | |
| XM_017012076.1 | 11 | 2719 | XP_016867565.1 | |
| XM_017012077.1 | 10 | 2573 | XP_016867566.1 | |
| XM_017012078.1 | 11 | 2598 | XP_016867567.1 | |
| XM_017012079.1 | 10 | 2521 | XP_016867568.1 | |
| XM_017012080.1 | 9 | 3232 | XP_016867569.1 | |
| AJ236921.1 | 10 | 3010 | CAB36968.1 | |
| AJ236922.1 | 11 | 3026 | CAB36969.1 | |
| BG202742.1 | 1 | 266 | ||
| BG211606.1 | 1 | 252 | ||
| U92459.1 | 10 | 2997 | AAB51764.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1027558 | Chr.7:126421217 - 126441119 on Build GRCh38 | Loss |
|
| esv3614931 | Chr.7:126334896 - 126534697 on Build GRCh38 | Loss |
|
| esv3891200 | Chr.7:124990785 - 126688176 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map