Assay Details
Target Gene Details
Entrez Gene ID: | 149041 |
Gene Name: | ring finger and CCCH-type domains 1 |
Gene Aliases: |
RNF198, ROQUIN |
Location: |
Chr.1:173931084-173993072 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RC3H1 | NM_001300850.1 | 19 | 5352 | NP_001287779.1 |
| NM_001300851.1 | 19 | 5325 | NP_001287780.1 | |
| NM_001300852.1 | 19 | 5322 | NP_001287781.1 | |
| NM_172071.3 | 19 | 5349 | NP_742068.1 | |
| XM_005244921.3 | 19 | 5289 | XP_005244978.1 | |
| XM_011509231.2 | 18 | 5178 | XP_011507533.1 | |
| AB095945.1 | 19 | 5190 | BAC23121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758978 | Chr.1:173841504 - 173986805 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map