Assay Details
Target Gene Details
Entrez Gene ID: | 10993 |
Gene Name: | serine dehydratase |
Gene Aliases: |
SDH |
Location: |
Chr.12:113392445-113403887 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDS | NM_006843.2 | 8 | 1064 | NP_006834.2 |
| AK124290.1 | 1 | 1254 | ||
| AK292760.1 | 8 | 1060 | ||
| BG566257.1 | 3 | 391 | ||
| BU740747.1 | 1 | 557 | ||
| J05037.1 | 8 | 1031 | AAA36604.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| esv3892265 | Chr.12:113099321 - 113527932 on Build GRCh38 | Gain |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map