Assay Details
Target Gene Details
Entrez Gene ID: | 84904 |
Gene Name: | Rho guanine nucleotide exchange factor 39 |
Gene Aliases: |
C9orf100 |
Location: |
Chr.9:35658875-35665281 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF39 | NM_032818.2 | 9 | 1541 | NP_116207.2 |
| XM_011518057.2 | 9 | 1542 | XP_011516359.1 | |
| XM_017015225.1 | 8 | 1413 | XP_016870714.1 | |
| XM_017015226.1 | 8 | 1336 | XP_016870715.1 | |
| XM_017015227.1 | 7 | 1188 | XP_016870716.1 | |
| AK001187.1 | 4 | 801 | ||
| AK027548.1 | 7 | 1093 | BAB55192.1 | |
| AK225501.1 | 9 | 1452 | ||
| AK225529.1 | 9 | 1456 | ||
| AY390225.1 | 9 | 1545 | AAR01010.1 | |
| AY390226.1 | 9 | 1545 | AAR01011.1 | |
| BC033666.1 | 9 | 1478 | AAH33666.1 |
More Information
Set Membership: |
|

Genomic Map