Assay Details
Target Gene Details
Entrez Gene ID: | 85397 |
Gene Name: | regulator of G-protein signaling 8 |
Gene Aliases: |
- |
Location: |
Chr.1:182641814-182753291 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3810 | Chr.1:182630610 - 182661590 on Build GRCh38 | Deletion |
|
| nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map