Assay Details
Target Gene Details
Entrez Gene ID: | 93973 |
Gene Name: | ARP8 actin-related protein 8 homolog |
Gene Aliases: |
ARP8, INO80N, hArp8 |
Location: |
Chr.3:53863096-53882202 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTR8 | NM_022899.4 | 13 | 1917 | NP_075050.3 |
| XM_005265587.4 | 13 | 1881 | XP_005265644.1 | |
| XM_011534249.2 | 13 | 1885 | XP_011532551.1 | |
| AK022996.1 | 13 | 1867 | BAB14352.1 | |
| AK026232.1 | 8 | 1070 | BAB15402.1 | |
| AK094507.1 | 13 | 1815 | ||
| BC032744.1 | 8 | 1088 | AAH32744.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1376e212 | Chr.3:53859682 - 53883597 on Build GRCh38 | Loss |
|
| nsv1013783 | Chr.3:53660391 - 53900754 on Build GRCh38 | Gain |
|
| esv3575570 | Chr.3:53721382 - 53883305 on Build GRCh38 | Gain |
|
| nsv1006990 | Chr.3:53815483 - 54251185 on Build GRCh38 | Gain |
|
| nsv1003621 | Chr.3:53371619 - 54391295 on Build GRCh38 | Loss |
|
| dgv4747n100 | Chr.3:53812714 - 53900754 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map