Assay Details
Target Gene Details
Entrez Gene ID: | 219844 |
Gene Name: | HYLS1, centriolar and ciliogenesis associated |
Gene Aliases: |
HLS |
Location: |
Chr.11:125883614-125900646 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HYLS1 | NM_001134793.1 | NP_001128265.1 | ||
| NM_145014.2 | NP_659451.1 | |||
| XM_005271430.2 | XP_005271487.1 | |||
| XM_006718777.3 | XP_006718840.1 | |||
| XM_011542659.2 | XP_011540961.1 | |||
| XM_017017320.1 | XP_016872809.1 | |||
| XM_017017321.1 | XP_016872810.1 | |||
| AK057477.1 | BAB71503.1 | |||
| AK127394.1 | ||||
| BC015047.1 | AAH15047.1 |
Target Gene Details
Entrez Gene ID: | 83480 |
Gene Name: | pseudouridylate synthase 3 |
Gene Aliases: |
2610020J05Rik, FKSG32 |
Location: |
Chr.11:125893485-125903774 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PUS3 | NM_001271985.1 | NP_001258914.1 | ||
| NM_031307.3 | NP_112597.3 | |||
| XM_005271687.3 | XP_005271744.1 | |||
| XM_005271688.3 | XP_005271745.1 | |||
| AF325689.1 | AAG50280.1 | |||
| AK055702.1 | BAB70990.1 | |||
| AK314251.1 | ||||
| BC004822.1 | AAH04822.1 | |||
| BM786445.1 | ||||
| CR982568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv248n67 | Chr.11:125843561 - 125919531 on Build GRCh38 | Gain |
|
| nsv468888 | Chr.11:125778321 - 125896149 on Build GRCh38 | Loss |
|
| nsv556525 | Chr.11:125690285 - 125971104 on Build GRCh38 | Gain |
|
| esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map