Assay Details
Target Gene Details
Entrez Gene ID: | 242 |
Gene Name: | arachidonate 12-lipoxygenase, 12R type |
Gene Aliases: |
12R-LOX, ARCI2 |
Location: |
Chr.17:8072636-8087703 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALOX12B | NM_001139.2 | 14 | 2125 | NP_001130.1 |
| AF038461.1 | 14 | 2123 | AAC39770.1 | |
| AF059250.1 | 14 | 1901 | AAC79680.1 | |
| BC041058.1 | 14 | 2153 | AAH41058.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953825 | Chr.17:8061683 - 8088382 on Build GRCh38 | Deletion |
|
| nsv833352 | Chr.17:8029719 - 8187575 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map