Assay Details
Target Gene Details
Entrez Gene ID: | 2260 |
Gene Name: | fibroblast growth factor receptor 1 |
Gene Aliases: |
BFGFR, CD331, CEK, ECCL, FGFBR, FGFR-1, FLG, FLT-2, FLT2, HBGFR, HH2, HRTFDS, KAL2, N-SAM, OGD, bFGF-R-1 |
Location: |
Chr.8:38411138-38468834 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951128 | Chr.8:38411783 - 38418282 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map