Assay Details
Target Gene Details
Entrez Gene ID: | 9056 |
Gene Name: | solute carrier family 7 member 7 |
Gene Aliases: |
LAT3, LPI, MOP-2, Y+LAT1, y+LAT-1 |
Location: |
Chr.14:22773222-22819811 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A7 | NM_001126105.2 | 4 | 824 | NP_001119577.1 |
| NM_001126106.2 | 4 | 930 | NP_001119578.1 | |
| NR_040448.1 | 3 | 1197 | ||
| XM_006720302.1 | 4 | 921 | XP_006720365.1 | |
| XM_011537298.2 | 3 | 832 | XP_011535600.1 | |
| XM_011537299.1 | 3 | 783 | XP_011535601.1 | |
| AB011263.1 | 2 | 582 | BAB11849.1 | |
| AB020532.1 | 3 | 1108 | BAA87623.1 | |
| AB209591.1 | 4 | 1991 | BAD92828.1 | |
| AF092032.1 | 4 | 868 | AAC83706.1 | |
| AJ130718.1 | 4 | 875 | CAA10198.1 | |
| AK025377.1 | 3 | 662 | ||
| AK301219.1 | 4 | 657 | ||
| AK309128.1 | 2 | 260 | ||
| AK314351.1 | 3 | 738 | ||
| AL365451.1 | 2 | 240 | ||
| BC003062.2 | 3 | 720 | AAH03062.1 | |
| BC010107.2 | 3 | 714 | AAH10107.1 | |
| BX161519.1 | 4 | 789 | CAD61952.1 | |
| BX248291.1 | 3 | 718 | CAD62619.1 | |
| KU178460.1 | 2 | 582 | ||
| KU178461.1 | ||||
| KU178462.1 | ||||
| Y18474.2 | 4 | 823 | CAB40136.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv127n21 | Chr.14:22690985 - 22790490 on Build GRCh38 | Gain |
|
| nsv563998 | Chr.14:22414183 - 22879743 on Build GRCh38 | Gain |
|
| dgv1836n100 | Chr.14:22676576 - 22803700 on Build GRCh38 | Gain |
|
| esv3633763 | Chr.14:22686795 - 22843098 on Build GRCh38 | Gain |
|
| nsv564039 | Chr.14:22629313 - 22794929 on Build GRCh38 | Gain |
|
| nsv819358 | Chr.14:22775794 - 22811167 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map