Assay Details
Target Gene Details
Entrez Gene ID: | 83879 |
Gene Name: | cell division cycle associated 7 |
Gene Aliases: |
ICF3, JPO1 |
Location: |
Chr.2:173354017-173368992 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDCA7 | NM_031942.4 | NP_114148.3 | ||
| NM_145810.2 | NP_665809.1 | |||
| XM_011511957.2 | XP_011510259.1 | |||
| AK027628.1 | BAB55245.1 | |||
| AK027642.1 | BAB55258.1 | |||
| AK075134.1 | 4 | 1465 | BAC11425.1 | |
| AK223524.1 | BAD97244.1 | |||
| AK297097.1 | ||||
| AK297245.1 | ||||
| AK300949.1 | ||||
| AL833728.1 | 1 | 134 | CAH56253.1 | |
| AL834186.1 | 4 | 1444 | CAH56357.1 | |
| AY029179.1 | AAK31591.1 | |||
| BC015124.1 | AAH15124.1 | |||
| BC027966.1 | AAH27966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834462 | Chr.2:173327577 - 173506798 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map