Assay Details
Target Gene Details
Entrez Gene ID: | 101927550 |
Gene Name: | uncharacterized LOC101927550 |
Gene Aliases: |
- |
Location: |
Chr.7:99013165-99036492 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927550 | NR_110102.1 | |||
| AK001533.1 | ||||
| DA099459.1 |
Target Gene Details
Entrez Gene ID: | 57154 |
Gene Name: | SMAD specific E3 ubiquitin protein ligase 1 |
Gene Aliases: |
- |
Location: |
Chr.7:99027435-99144120 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMURF1 | NM_001199847.1 | NP_001186776.1 | ||
| NM_020429.2 | NP_065162.1 | |||
| NM_181349.2 | NP_851994.1 | |||
| XM_017012457.1 | XP_016867946.1 | |||
| AB046845.1 | ||||
| AF199364.2 | AAF08298.2 | |||
| AF464850.1 | AAM90910.1 | |||
| AL834242.1 | CAD38919.1 | |||
| BC136804.1 | ||||
| BC144414.1 | ||||
| BC152468.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3614267 | Chr.7:99027640 - 99036461 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map