Assay Details
Target Gene Details
Entrez Gene ID: | 120425 |
Gene Name: | junction adhesion molecule like |
Gene Aliases: |
AMICA, AMICA1, CREA7-1, CREA7-4, Gm638 |
Location: |
Chr.11:118193727-118225094 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAML | NM_001098526.1 | 7 | 1044 | NP_001091996.1 |
| NM_001286570.1 | 6 | 981 | NP_001273499.1 | |
| NM_001286571.1 | 6 | 1272 | NP_001273500.1 | |
| NM_153206.2 | 6 | 1282 | NP_694938.2 | |
| NR_104479.1 | 6 | 1272 | ||
| XM_005271407.3 | 6 | 982 | XP_005271464.1 | |
| XM_017017213.1 | 7 | 969 | XP_016872702.1 | |
| XM_017017214.1 | 6 | 929 | XP_016872703.1 | |
| XM_017017215.1 | 5 | 857 | XP_016872704.1 | |
| AJ515553.2 | 7 | 964 | CAD56620.2 | |
| AK090409.1 | 6 | 3759 | BAC03390.1 | |
| AK094399.1 | 6 | 1282 | BAC04347.1 | |
| AK097285.1 | 2 | 907 | ||
| AK298722.1 | 6 | 898 | ||
| AK300851.1 | 6 | 592 | ||
| AK301099.1 | 7 | 961 | ||
| AW804077.1 | 1 | 51 | ||
| AY093686.1 | 7 | 961 | AAM15730.1 | |
| AY093687.1 | 5 | 753 | AAM15731.1 | |
| AY138965.1 | 7 | 945 | AAN52117.1 | |
| AY358362.1 | 7 | 954 | AAQ88728.1 | |
| BC100794.2 | 6 | 971 | AAI00795.1 | |
| BC100795.2 | 6 | 971 | AAI00796.1 | |
| BC100796.1 | 6 | 971 | AAI00797.1 | |
| BC100797.1 | 6 | 971 | AAI00798.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3579923 | Chr.11:118193687 - 118204244 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map