Assay Details
Target Gene Details
Entrez Gene ID: | 149095 |
Gene Name: | DC-STAMP domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.1:155033806-155050930 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCST1 | NM_001143687.2 | NP_001137159.1 | ||
| NM_152494.3 | NP_689707.2 | |||
| AK057347.1 | BAB71440.1 | |||
| AK301876.1 | ||||
| AK301902.1 | ||||
| AK301935.1 | ||||
| BC064844.1 | AAH64844.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
| nsv952212 | Chr.1:154998225 - 155087624 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map