Assay Details
Target Gene Details
Entrez Gene ID: | 7314 |
Gene Name: | ubiquitin B |
Gene Aliases: |
HEL-S-50 |
Location: |
Chr.17:16380793-16382745 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBB | NM_001281716.1 | NP_001268645.1 | ||
| NM_001281720.1 | NP_001268649.1 | |||
| NM_018955.3 | 1 | 267 | NP_061828.1 | |
| AK129498.1 | ||||
| AK300882.1 | ||||
| BC000379.2 | AAH00379.1 | |||
| BC015127.1 | AAH15127.1 | |||
| BC026301.1 | AAH26301.1 | |||
| BC031027.1 | AAH31027.1 | |||
| BC038999.1 | AAH38999.1 | |||
| BC046123.2 | AAH46123.1 | |||
| BP307496.1 | 1 | 267 | ||
| BU661443.1 | ||||
| GQ472200.1 | ||||
| HY014043.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063764 | Chr.17:16268335 - 16759600 on Build GRCh38 | Loss |
|
| esv2715702 | Chr.17:15750514 - 16668268 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map