Assay Details
Target Gene Details
Entrez Gene ID: | 131601 |
Gene Name: | transmembrane protein adipocyte associated 1 |
Gene Aliases: |
GPR175, TMEM227, TPRA40 |
Location: |
Chr.3:127573064-127598234 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPRA1 | NM_001136053.2 | 11 | 1711 | NP_001129525.1 |
| NM_001142646.2 | 9 | 1547 | NP_001136118.1 | |
| NR_073377.1 | 10 | 1650 | ||
| XM_006713495.3 | 12 | 1892 | XP_006713558.1 | |
| XM_006713496.2 | 11 | 1569 | XP_006713559.1 | |
| XM_006713497.3 | 12 | 1936 | XP_006713560.1 | |
| XM_011512422.1 | 11 | 1485 | XP_011510724.1 | |
| XM_017005707.1 | 12 | 1675 | XP_016861196.1 | |
| XM_017005708.1 | 12 | 1705 | XP_016861197.1 | |
| XM_017005709.1 | 11 | 1745 | XP_016861198.1 | |
| XM_017005710.1 | 11 | 1584 | XP_016861199.1 | |
| AB037108.1 | 10 | 1635 | BAA89782.1 | |
| AF289568.1 | 9 | 1494 | AAL55752.1 | |
| AI248453.1 | 1 | 239 | ||
| AK056759.1 | 1 | 1721 | ||
| BC017058.1 | 10 | 1419 | AAH17058.1 | |
| BC050711.2 | 11 | 1605 | AAH50711.1 | |
| BM665705.1 | 1 | 254 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8563n54 | Chr.3:127543914 - 127685471 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map