Assay Details
Target Gene Details
Entrez Gene ID: | 533 |
Gene Name: | ATPase H+ transporting V0 subunit b |
Gene Aliases: |
ATP6F, HATPL, VMA16 |
Location: |
Chr.1:43974648-43978300 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6V0B | NM_001039457.2 | NP_001034546.1 | ||
| NM_001294333.1 | NP_001281262.1 | |||
| NM_004047.4 | NP_004038.1 | |||
| AI439545.1 | ||||
| BC000423.2 | AAH00423.1 | |||
| BC005876.2 | AAH05876.1 | |||
| BG475609.1 | ||||
| BG831921.1 | ||||
| BI770034.1 | ||||
| BT007151.1 | AAP35815.1 | |||
| CR456972.1 | CAG33253.1 | |||
| D89052.1 | BAA13753.1 | |||
| L05089.1 | AAC15853.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546141 | Chr.1:43975861 - 43985562 on Build GRCh38 | Loss |
|
| nsv732 | Chr.1:43976690 - 43992799 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map