Assay Details
Target Gene Details
Entrez Gene ID: | 57447 |
Gene Name: | NDRG family member 2 |
Gene Aliases: |
SYLD |
Location: |
Chr.14:21016763-21070872 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDRG2 | NM_001282211.1 | NP_001269140.1 | ||
| AK293514.1 |
Target Gene Details
Entrez Gene ID: | 84659 |
Gene Name: | ribonuclease A family member 7 |
Gene Aliases: |
RAE1 |
Location: |
Chr.14:21042226-21044233 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE7 | NM_032572.3 | 2 | 761 | NP_115961.2 |
| AJ131212.1 | 1 | 504 | CAC20410.1 | |
| AJ306608.1 | 2 | 671 | CAC84462.1 | |
| AJ306609.1 | 2 | 671 | CAC84457.1 | |
| AJ306610.1 | 2 | 671 | CAC84458.1 | |
| AK292697.1 | 2 | 761 | ||
| AY170392.1 | 2 | 735 | AAO12510.1 | |
| AY359097.1 | 2 | 603 | AAQ89455.1 | |
| BC074960.2 | AAH74960.1 | |||
| BC112334.1 | 2 | 599 | AAI12335.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428293 | Chr.14:20747949 - 21063624 on Build GRCh38 | Gain+Loss |
|
| nsv832740 | Chr.14:20863901 - 21076893 on Build GRCh38 | Gain |
|
| nsv826882 | Chr.14:21042862 - 21136042 on Build GRCh38 | Gain |
|
| nsv1039369 | Chr.14:21030895 - 21062142 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map