Assay Details
Target Gene Details
Entrez Gene ID: | 8816 |
Gene Name: | DDB1 and CUL4 associated factor 5 |
Gene Aliases: |
BCRG2, BCRP2, D14S1461E, WDR22 |
Location: |
Chr.14:69050920-69153393 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF5 | NM_001284206.1 | 9 | 2952 | NP_001271135.1 |
| NM_001284207.1 | 9 | 2708 | NP_001271136.1 | |
| NM_003861.2 | 9 | 2955 | NP_003852.1 | |
| XM_006720297.2 | 10 | 3060 | XP_006720360.1 | |
| XM_006720298.2 | 10 | 3057 | XP_006720361.1 | |
| XM_006720299.3 | 10 | 2695 | XP_006720362.1 | |
| XM_006720300.2 | 10 | 2716 | XP_006720363.1 | |
| XM_011537278.1 | 11 | 3329 | XP_011535580.1 | |
| XM_011537279.2 | 10 | 2792 | XP_011535581.1 | |
| XM_011537280.2 | 6 | 2283 | XP_011535582.1 | |
| XM_017021733.1 | 10 | 2650 | XP_016877222.1 | |
| XM_017021734.1 | 9 | 2607 | XP_016877223.1 | |
| XM_017021735.1 | 9 | 2687 | XP_016877224.1 | |
| XM_017021736.1 | 9 | 2611 | XP_016877225.1 | |
| XM_017021737.1 | 5 | 2240 | XP_016877226.1 | |
| XM_017021738.1 | 10 | 3772 | XP_016877227.1 | |
| XM_017021739.1 | 10 | 3859 | XP_016877228.1 | |
| AB058727.1 | 9 | 2790 | ||
| AF044774.1 | 4 | 1953 | AAC08965.1 | |
| AL831823.2 | 1 | 1467 | CAD38537.2 | |
| AL831932.2 | 9 | 2708 | CAD38589.1 | |
| AL834533.1 | 1 | 568 | CAD39189.1 | |
| BC136632.1 | 9 | 2884 | ||
| BX648006.1 | 9 | 2759 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952860 | Chr.14:69053584 - 69055983 on Build GRCh38 | Deletion |
|
| nsv832823 | Chr.14:68922136 - 69100921 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map