Assay Details
Target Gene Details
Entrez Gene ID: | 84285 |
Gene Name: | eukaryotic translation initiation factor 1A domain containing |
Gene Aliases: |
haponin |
Location: |
Chr.11:65996545-66002217 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF1AD | NM_001242481.1 | 6 | 1120 | NP_001229410.1 |
| NM_001242482.1 | 6 | 1096 | NP_001229411.1 | |
| NM_001242483.1 | 6 | 1021 | NP_001229412.1 | |
| NM_001242484.1 | 6 | 990 | NP_001229413.1 | |
| NM_001242485.1 | 6 | 966 | NP_001229414.1 | |
| NM_001242486.1 | 6 | 891 | NP_001229415.1 | |
| NM_032325.3 | 6 | 1091 | NP_115701.2 | |
| XM_017018412.1 | 6 | 1252 | XP_016873901.1 | |
| AK094129.1 | 6 | 1032 | BAC04293.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832191 | Chr.11:65864858 - 66038649 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map