Assay Details
Target Gene Details
Entrez Gene ID: | 347516 |
Gene Name: | diacylglycerol O-acyltransferase 2 like 6 |
Gene Aliases: |
DC3 |
Location: |
Chr.X:70177483-70205703 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DGAT2L6 | NM_198512.2 | NP_940914.1 | ||
| AK129500.1 | BAC85167.1 | |||
| BG696880.1 |
More Information
Set Membership: |
|

Genomic Map