Assay Details
Target Gene Details
Entrez Gene ID: | 10058 |
Gene Name: | ATP binding cassette subfamily B member 6 (Langereis blood group) |
Gene Aliases: |
ABC, ABC14, DUH3, LAN, MCOPCB7, MTABC3, PRP, PSHK2, umat |
Location: |
Chr.2:219209766-219218990 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCB6 | NM_005689.2 | 19 | 2938 | NP_005680.1 |
| AF070598.1 | 15 | 1630 | AAC28653.1 | |
| AF308472.1 | 19 | 2927 | AAG33617.1 | |
| AJ289233.2 | 19 | 2898 | CAB95766.2 | |
| AK026067.1 | 16 | 1747 | ||
| AK057026.1 | 19 | 2657 | BAB71347.1 | |
| AK172812.1 | 19 | 2288 | BAD18782.1 | |
| AW664349.1 | ||||
| BC000559.2 | 19 | 2889 | AAH00559.1 | |
| BC043423.1 | 12 | 2938 | AAH43423.1 | |
| U66673.1 | 8 | 920 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893555 | Chr.2:219172722 - 219273269 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map