Assay Details
Target Gene Details
Entrez Gene ID: | 27293 |
Gene Name: | sphingomyelin phosphodiesterase acid like 3B |
Gene Aliases: |
ASML3B |
Location: |
Chr.1:27934955-27959157 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMPDL3B | NM_001009568.2 | 1 | 102 | NP_001009568.1 |
| NM_001304579.1 | 1 | 102 | NP_001291508.1 | |
| NM_014474.3 | 1 | 102 | NP_055289.2 | |
| XM_011541259.2 | 1 | 102 | XP_011539561.1 | |
| AK293489.1 | ||||
| AK293566.1 | ||||
| AK301349.1 | 1 | 57 | ||
| AK316500.1 | ||||
| BC014444.1 | AAH14444.1 | |||
| BP284513.1 | 1 | 102 | ||
| CB160850.1 | 1 | 64 | ||
| DC370191.1 | 1 | 101 | ||
| Y08134.1 | CAA69328.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585564 | Chr.1:27879549 - 27935475 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map