Assay Details
Target Gene Details
Entrez Gene ID: | 420 |
Gene Name: | ADP-ribosyltransferase 4 (Dombrock blood group) |
Gene Aliases: |
ARTC4, CD297, DO, DOK1 |
Location: |
Chr.12:14829311-14843479 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ART4 | NM_021071.2 | NP_066549.2 | ||
| AK291662.1 | ||||
| AY829433.1 | AAX39380.1 | |||
| AY829434.1 | AAX39381.1 | |||
| CR746266.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3437825 | Chr.12:14843265 - 14843775 on Build GRCh38 | Duplication |
|
| esv3302544 | Chr.12:14843254 - 14843766 on Build GRCh38 | Tandem Duplication |
|
More Information
Set Membership: |
|

Genomic Map