Assay Details
Target Gene Details
Entrez Gene ID: | 11218 |
Gene Name: | DEAD-box helicase 20 |
Gene Aliases: |
DP103, GEMIN3 |
Location: |
Chr.1:111755568-111767577 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX20 | NM_007204.4 | 1 | 260 | NP_009135.4 |
| AF171063.1 | 1 | 196 | AAF14544.1 | |
| BC034953.2 | 1 | 231 | AAH34953.1 | |
| DB072947.1 |
Target Gene Details
Entrez Gene ID: | 55924 |
Gene Name: | family with sequence similarity 212 member B |
Gene Aliases: |
C1orf183 |
Location: |
Chr.1:111722064-111755797 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM212B | NM_198926.2 | NP_945120.1 | ||
| AK293989.1 | ||||
| AL137199.1 | CAB69909.1 |
More Information
Set Membership: |
|

Genomic Map