Assay Details
Target Gene Details
Entrez Gene ID: | 63895 |
Gene Name: | piezo type mechanosensitive ion channel component 2 |
Gene Aliases: |
C18orf30, C18orf58, DA3, DA5, FAM38B, FAM38B2, HsT748, HsT771, MWKS |
Location: |
Chr.18:10670237-11149561 on Build GRCh38 |
Assay Gene Location: | Within Exon 55 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIEZO2 | NM_022068.3 | 52 | 8397 | NP_071351.2 |
| XM_011525723.2 | 55 | 9327 | XP_011524025.1 | |
| XM_011525724.2 | 54 | 9270 | XP_011524026.1 | |
| XM_011525725.2 | 53 | 9237 | XP_011524027.1 | |
| XM_011525726.2 | 54 | 9144 | XP_011524028.1 | |
| XM_017025918.1 | 54 | 9288 | XP_016881407.1 | |
| AK026797.1 | 4 | 558 | BAB15556.1 | |
| AK027056.1 | 11 | 1615 | BAB15641.1 | |
| AK302764.1 | 14 | 2234 | ||
| JN790819.1 | 52 | 8231 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057995 | Chr.18:10626172 - 11923368 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map