Assay Details
Target Gene Details
Entrez Gene ID: | 1728 |
Gene Name: | NAD(P)H quinone dehydrogenase 1 |
Gene Aliases: |
DHQU, DIA4, DTD, NMOR1, NMORI, QR1 |
Location: |
Chr.16:69709401-69726668 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NQO1 | NM_000903.2 | NP_000894.1 | ||
| NM_001025433.1 | NP_001020604.1 | |||
| NM_001025434.1 | NP_001020605.1 | |||
| NM_001286137.1 | NP_001273066.1 | |||
| AK223050.1 | BAD96770.1 | |||
| AK297125.1 | ||||
| AK297979.1 | ||||
| AK298896.1 | ||||
| AK307533.1 | ||||
| AK309513.1 | ||||
| AK312368.1 | ||||
| AK316246.1 | ||||
| BC007659.2 | AAH07659.1 | |||
| BC107739.1 | AAI07740.1 | |||
| BE903613.1 | ||||
| CD014005.1 | ||||
| J03934.1 | AAA59940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524364 | Chr.16:69711242 - 69769540 on Build GRCh38 | Loss |
|
| esv3892878 | Chr.16:69687512 - 69844258 on Build GRCh38 | Gain |
|
| esv3638928 | Chr.16:69677077 - 69756070 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map