Assay Details
Target Gene Details
Entrez Gene ID: | 57704 |
Gene Name: | glucosylceramidase beta 2 |
Gene Aliases: |
AD035, NLGase, SPG46 |
Location: |
Chr.9:35736862-35749255 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBA2 | NM_020944.2 | NP_065995.1 | ||
| XM_005251526.4 | XP_005251583.1 | |||
| XM_006716809.3 | XP_006716872.1 | |||
| XM_011517973.2 | XP_011516275.1 | |||
| XM_017014937.1 | XP_016870426.1 | |||
| XM_017014938.1 | XP_016870427.1 | |||
| XM_017014939.1 | XP_016870428.1 | |||
| XM_017014940.1 | XP_016870429.1 | |||
| XM_017014941.1 | XP_016870430.1 | |||
| XM_017014942.1 | XP_016870431.1 | |||
| XM_017014943.1 | XP_016870432.1 | |||
| XM_017014944.1 | XP_016870433.1 | |||
| XM_017014945.1 | XP_016870434.1 | |||
| XM_017014946.1 | XP_016870435.1 | |||
| AB046825.1 | BAB13431.1 | |||
| AF258662.1 | AAG44660.1 | |||
| AJ309567.1 | CAC83792.1 | |||
| AK027884.1 | BAB55430.1 | |||
| AK057610.1 | 1 | 277 | ||
| AK295967.1 | ||||
| AK297435.1 | ||||
| AL834306.1 | CAD38976.1 | |||
| BC011363.1 | AAH11363.1 |
More Information
Set Membership: |
|

Genomic Map