Assay Details
Target Gene Details
Entrez Gene ID: | 3572 |
Gene Name: | interleukin 6 signal transducer |
Gene Aliases: |
CD130, CDW130, GP130, IL-6RB |
Location: |
Chr.5:55935095-55994993 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL6ST | NM_001190981.1 | NP_001177910.1 | ||
| NM_002184.3 | NP_002175.2 | |||
| NM_175767.2 | NP_786943.1 | |||
| NR_120480.1 | ||||
| XM_017009441.1 | XP_016864930.1 | |||
| XM_017009442.1 | XP_016864931.1 | |||
| AB015706.1 | BAA78112.1 | |||
| AB102801.1 | BAD89392.1 | |||
| AB102802.1 | BAD89393.1 | |||
| AB102804.1 | BAD89395.1 | |||
| BC117402.1 | ||||
| BC117404.1 | ||||
| HQ628629.1 | ||||
| HQ628630.1 | ||||
| M57230.1 | AAA59155.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830307 | Chr.5:55867930 - 56048443 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map