Assay Details
Target Gene Details
Entrez Gene ID: | 339221 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 7 |
Gene Aliases: |
ALK-SMase, E-NPP 7, NPP-7, NPP7 |
Location: |
Chr.17:79730919-79742926 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENPP7 | NM_178543.4 | 2 | 394 | NP_848638.3 |
| XM_011524737.1 | 2 | 578 | XP_011523039.1 | |
| AK126250.1 | 2 | 578 | BAC86504.1 | |
| AY230663.1 | 2 | 377 | AAP69661.1 | |
| AY358622.1 | 2 | 360 | AAQ88985.1 | |
| BC041453.1 | 2 | 377 | AAH41453.2 | |
| DB187443.1 | 2 | 394 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952381 | Chr.17:79677839 - 79741901 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map