Assay Details
Target Gene Details
Entrez Gene ID: | 25778 |
Gene Name: | dual serine/threonine and tyrosine protein kinase |
Gene Aliases: |
CAKUT1, DustyPK, HDCMD38P, RIP5, RIPK5 |
Location: |
Chr.1:205142497-205211599 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSTYK | NM_015375.2 | 13 | 6677 | NP_056190.1 |
| NM_199462.2 | 12 | 6542 | NP_955749.1 | |
| XM_011509392.2 | 13 | 6608 | XP_011507694.1 | |
| XM_011509393.2 | 14 | 6571 | XP_011507695.1 | |
| XM_011509394.2 | 12 | 6079 | XP_011507696.1 | |
| AB007941.1 | 8 | 4923 | BAA32317.1 | |
| AY429674.1 | 13 | 6649 | AAS55390.1 | |
| BU168830.1 | 1 | 169 | ||
| BU170290.1 | 1 | 283 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv563n100 | Chr.1:205101394 - 205152107 on Build GRCh38 | Gain |
|
| nsv549042 | Chr.1:205114992 - 205193929 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map