Assay Details
Target Gene Details
Entrez Gene ID: | 4649 |
Gene Name: | myosin IXA |
Gene Aliases: |
- |
Location: |
Chr.15:71822290-72118577 on Build GRCh38 |
Assay Gene Location: | Within Exon 48 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYO9A | NM_006901.3 | 42 | 7964 | NP_008832.2 |
| XM_006720539.2 | 43 | 8177 | XP_006720602.1 | |
| XM_011521613.2 | 44 | 8234 | XP_011519915.1 | |
| XM_011521614.2 | 45 | 7920 | XP_011519916.1 | |
| XM_011521615.2 | 44 | 8087 | XP_011519917.1 | |
| XM_011521616.2 | 44 | 7881 | XP_011519918.1 | |
| XM_011521617.2 | 44 | 8231 | XP_011519919.1 | |
| XM_011521618.2 | 43 | 8180 | XP_011519920.1 | |
| XM_011521619.2 | 43 | 8177 | XP_011519921.1 | |
| XM_011521620.2 | 43 | 8174 | XP_011519922.1 | |
| XM_011521621.2 | 43 | 8114 | XP_011519923.1 | |
| XM_011521622.2 | 43 | 8018 | XP_011519924.1 | |
| XM_011521623.2 | 27 | 5337 | XP_011519925.1 | |
| XM_017022228.1 | 44 | 7863 | XP_016877717.1 | |
| XM_017022229.1 | 44 | 7853 | XP_016877718.1 | |
| XM_017022230.1 | 42 | 7961 | XP_016877719.1 | |
| AB290183.1 | 41 | 7399 | ||
| AF117888.1 | 42 | 7715 | AAD49195.1 | |
| AK001923.1 | 11 | 1525 | BAA91979.1 | |
| AL137287.1 | 19 | 4022 | CAB70679.1 | |
| BC140869.1 | 42 | 7633 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892712 | Chr.15:71809756 - 72033254 on Build GRCh38 | Loss |
|
| nsv1036938 | Chr.15:71655767 - 71980514 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map