Assay Details
Target Gene Details
Entrez Gene ID: | 79813 |
Gene Name: | euchromatic histone lysine methyltransferase 1 |
Gene Aliases: |
EUHMTASE1, Eu-HMTase1, FP13812, GLP, GLP1, KMT1D, bA188C12.1 |
Location: |
Chr.9:137618988-137836127 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Target Gene Details
Entrez Gene ID: | 643210 |
Gene Name: | EHMT1 intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.9:137763022-137764772 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EHMT1-IT1 | NR_024341.1 | 1 | 829 | |
| AK097611.1 | 1 | 829 | BAC05120.1 | |
| AK127990.1 | 1 | 829 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509337 | Chr.9:137756805 - 137794033 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map