Assay Details
Target Gene Details
Entrez Gene ID: | 1937 |
Gene Name: | eukaryotic translation elongation factor 1 gamma |
Gene Aliases: |
EF1G, GIG35 |
Location: |
Chr.11:62559601-62573988 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EEF1G | NM_001404.4 | NP_001395.1 | ||
| AK092787.1 | ||||
| AK129569.1 | ||||
| AK129618.1 | ||||
| AK130026.1 | ||||
| AK299876.1 | ||||
| AK300203.1 | ||||
| AK300686.1 | ||||
| AK300729.1 | ||||
| AY542307.1 | AAT08176.1 | |||
| M55409.1 | AAC18414.1 | |||
| X63526.1 | CAA45089.1 | |||
| Z11531.1 | CAA77630.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584599 | Chr.11:62495270 - 62630578 on Build GRCh38 | Gain |
|
| nsv832180 | Chr.11:62434133 - 62597356 on Build GRCh38 | Loss |
|
| nsv347 | Chr.11:62529596 - 62571455 on Build GRCh38 | Deletion |
|
| nsv825939 | Chr.11:62558611 - 62643105 on Build GRCh38 | Gain |
|
| nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss |
|
| nsv975193 | Chr.11:62559450 - 62560565 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map