Assay Details
Target Gene Details
Entrez Gene ID: | 4644 |
Gene Name: | myosin VA |
Gene Aliases: |
GS1, MYH12, MYO5, MYR12 |
Location: |
Chr.15:52307283-52529050 on Build GRCh38 |
Assay Gene Location: | Within Exon 43 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYO5A | NM_000259.3 | 41 | 10381 | NP_000250.3 |
| NM_001142495.1 | 40 | 10300 | NP_001135967.1 | |
| XM_011521606.2 | 42 | 10291 | XP_011519908.2 | |
| AB209505.1 | 33 | 8913 | BAD92742.1 | |
| AB290180.1 | 40 | 10059 | ||
| AK090859.1 | 1 | 184 | ||
| AK124898.1 | 1 | 2812 |
More Information
Set Membership: |
|

Genomic Map